Dunkar, rektangulära, med kran, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Made of FLPE with TFE spigot and fluorinated PP screw cap.
- Graduated, tough rigid and translucent
- Space-saving rectangular shape
- Attached stainless steel handle
Excellent for storing and dispensing a variety of solvents; useful for collecting and dispensing distilled water. Fluorinated surface (inside and outside) improves barrier properties; reduces solvent absorption and permeation.
Specifications
- Catalog No:
- 216-1222
- 216-1223
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Miljöcertificering:
- NAR Certificate of Retirement
- NAR Certificate of Retirement