Orders LinkContactUsLinkComponent
Dunkar, Biotainer®, Nalgene®
Dunkar, Biotainer®, Nalgene®
Dunkar, Biotainer®, Nalgene®
Dunkar, Biotainer®, Nalgene®
PC, clear, with white screw cap.
  • Silicone-lined polypropylene screw cap
  • Good chemical resistance
  • Good temperature resistance
Reduce the risk of carry-over contamination and eliminate the cost of cleaning, sterilizing and associated validations. Use sterile, ready-to-use Biotainer™ Carboys, designed specifically for pharmaceutical and biological manufacturers. Excellent for freezing applications.
  • Catalog No:
  • 391-7103
  • 391-7102
  • 391-7104
  • 391-7100
  • 391-7101
  • NALG3405-99
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Miljöcertificering:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Product Family Options
  • Kapacitet
    Typ
    Ø utv.
    Hals, innerdiam.
    Höjd
    Gänga
  • 10 l
    med handtag, styckförpackade
    250 mm
    37 mm
    337 mm
    48 mm
    1 * 2 Items
    1 * 2 Items - 2 325,00 kr
    2 325,00 kr
    1 * 2 Items
       2 325,00 kr
      Low Manufacturing Impact
    Specifications
    Cat. No.
    391-7103
    Environmentally Preferable
    Low Manufacturing Impact
    Miljöcertificering
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 10 l
    med handtag, bulkförpackade
    250 mm
    37 mm
    337 mm
    48 mm
    1 * 2 Items
    1 * 2 Items - 3 625,00 kr
    3 625,00 kr
    1 * 2 Items
       3 625,00 kr
      Low Manufacturing Impact
    Specifications
    Cat. No.
    391-7102
    Environmentally Preferable
    Low Manufacturing Impact
    Miljöcertificering
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 20 l
    med handtag, styckförpackade
    255 mm
    37 mm
    494 mm
    48 mm
    1 * 3 Items
    1 * 3 Items - 4 600,00 kr
    4 600,00 kr
    1 * 3 Items
       4 600,00 kr
      Low Manufacturing Impact
    Specifications
    Cat. No.
    391-7104
    Environmentally Preferable
    Low Manufacturing Impact
    Miljöcertificering
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 5 l
    med handtag, styckförpackade
    166 mm
    38 mm
    290 mm
    48 mm
    1 * 6 Items
    1 * 6 Items - 5 850,00 kr
    5 850,00 kr
    1 * 6 Items
       5 850,00 kr
      Low Manufacturing Impact
    Specifications
    Cat. No.
    391-7100
    Environmentally Preferable
    Low Manufacturing Impact
    Miljöcertificering
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 5 l
    utan handtag, styckförpackade
    166 mm
    38 mm
    290 mm
    48 mm
    1 * 6 Items
    1 * 6 Items - 4 875,00 kr
    4 875,00 kr
    1 * 6 Items
       4 875,00 kr
      Low Manufacturing Impact
    Specifications
    Cat. No.
    391-7101
    Environmentally Preferable
    Low Manufacturing Impact
    Miljöcertificering
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 5 l
    1 * 6 Items
    1 * 6 Items
    Specifications
    Cat. No.
    NALG3405-99
    Environmentally Preferable
    Miljöcertificering