ContactUsLinkComponent
Bägare, låg modell, PTFE
Bägare, låg modell, PTFE
Katalog # 213-1556
Leverantör:  Brand
Leverantör #: 90328
undefined
Bägare, låg modell, PTFE
Katalog # 213-1556
Leverantör:  Brand
Leverantör #: 90328
Logga in för tillgänglighet
Select an Option
43 mm
Select an Option
50 ml
Select an Option
55 mm
Select an Option
04
Select an Option
1
Om denna produkt
PTFE, with spout.
  • Excellent chemical resistance
  • Temperature resistant from −200 to +280 °C
  • Flat bottom for good heat transfer, for example when used on regulated hotplates
Thick walled, with reinforced lip and exceptionally smooth inner walls to prevent contamination. Non stick. Low gas permeability.