Polypropylene microplates, 96- and 384-well
Katalog # 737-0237
Leverantör: EPPENDORF
Specifikationer
- Förp.:12×20
- Steril:-
- Färg:Klar
- Beskrivning:Microplates 384/V, white border, PCR clean, LoBind®
- Seq:20
- Environmentally Preferable:Low Manufacturing Impact
- Miljöcertificering:Renewable Energy Certificate, ISO 14001
- Arbetsvolym:5 - 120 µl
- Total volym:140 µl
Specifikationer
Om denna produkt
These polypropylene microplates feature high transparency and easy to read alphanumeric labelling, making the identification of the correct plate or an individual well quick, easy, and less error-prone. Black and white versions are available for fluorescence or luminescence detection, offering excellent signal-to-noise ratios for improved sensitivity.
- Highest transparency for better sample visibility
- OptiTrack® matrix for easy and fast well identification
- g-safe® for time saving centrifugation up to 6000×g
- Well designed for maximum sample recovery with raised rim for effective sealing
- High chemical resistance
All Eppendorf microplates are manufactured in ISO certified clean room conditions. They are available in the purity levels PCR clean (free of human DNA, DNase, RNase and PCR inhibitors) or sterile. Each batch is tested and certified individually.