Fibrillar Collagen 24-Well Multiwell Insert System, Corning® BioCoat®
Catalog # 734-1079
Supplier: Corning
Specifications
- Membran:PET
- Porstorlek:1,0 µm
- Beskrivning:Fibrillar collagen 24-multiwell inserts, with feeder tray and lid
- Environmentally Preferable:Low Manufacturing Impact
- Miljöcertificering:ISO 14001 - Bedford
- Membrandiameter:6,4 mm
- Förp.:1
Specifications
About this item
This system is packaged with ready-to-use Falcon® Companion Plates.