Renrumshandskar, nitril, sterila, Kimtech™ G3 Sterling™ Sterile
Supplier: KIMBERLY CLARK
Certificates
About this item
Hand-specific, sterile gloves with beaded cuff for cleanroom applications. When double donned, these gloves deliver the equivalent feel and dexterity of a single latex glove without the risk of type I latex allergic reactions.
- Sterile, powder-free, textured finish
- Environmentally responsible thanks to improved packaging that saves waste
- Static dissipative in use
- Comfortable fit, suitable for double donning
- Particles per cm² ≥0,5 micron: max. 1200
Suitable for use in Class 1, ISO 3 or M1.5 or higher cleanrooms.
Length: 300 mm
Thickness: 0,08 mm (palm)
Colour: Grey
Certifications: PPE Cat III (EU) 2016/425, EN 374-1 Type C (K), EN 374-5 (VIRUS), AQL 1.5
Packaging: Double packed in polyethylene. 30 pairs per bag.
Specifications
- Catalog No:
- 112-1864
- 112-1865
- 112-1862
- 112-1863
- 112-1861
- 112-1858
- 112-1859
- 112-1860
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Environmentally Preferable:
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Waste Reducing
- Engångs/Återanvändbar:
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Latex/latexfri:
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Pudrad/puderfri:
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Renrumsklass:
- 1/ISO 3
- 1/ISO 3
- 1/ISO 3
- 1/ISO 3
- 1/ISO 3
- 1/ISO 3
- 1/ISO 3
- 1/ISO 3
- Kragmodell:
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Krage, tjocklek:
- 0,07 mm
- 0,07 mm
- 0,07 mm
- 0,07 mm
- 0,07 mm
- 0,07 mm
- 0,07 mm
- 0,07 mm
- Finger, tjocklek:
- 0,10 mm
- 0,10 mm
- 0,10 mm
- 0,10 mm
- 0,10 mm
- 0,10 mm
- 0,10 mm
- 0,10 mm
- Handflata, tjocklek:
- 0,08 mm
- 0,08 mm
- 0,08 mm
- 0,08 mm
- 0,08 mm
- 0,08 mm
- 0,08 mm
- 0,08 mm
- EN standard:
- EN 374-1 Type C (K), EN 374-5 (VIRUS)
- EN 374-1 Type C (K), EN 374-5 (VIRUS)
- EN 374-1 Type C (K), EN 374-5 (VIRUS)
- EN 374-1 Type C (K), EN 374-5 (VIRUS)
- EN 374-1 Type C (K), EN 374-5 (VIRUS)
- EN 374-1 Type C (K), EN 374-5 (VIRUS)
- EN 374-1 Type C (K), EN 374-5 (VIRUS)
- EN 374-1 Type C (K), EN 374-5 (VIRUS)
- Antistatisk:
- +
- +
- +
- +
- +
- +
- +
- +