Orders LinkContactUsLinkComponent
Sampler, UniSampler “Ex”
Sampler, UniSampler “Ex”
UniSampler "Ex"
Katalog # 300-1044
Leverantör:   BURKLE
Sampler, UniSampler “Ex”
Katalog # 300-1044
Leverantör:  BURKLE
Sampler, UniSampler “Ex”
Sampler, UniSampler “Ex”
UniSampler "Ex"
Price
7 300,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Produktdetaljer och dokument
Glass bottle and hose are connected in such a way that they conduct electricity and are grounded via a grounding cable. Operation: Ground the device, immerse the suction hose to the required depth, attach the adapter to the sampling bottle, generate a vacuum with the manual pump and collect the sample, remove the filled sampling bottle, close it and label it.
  • Specially developed for sampling flammable liquids of hazardous materials (e.g. gasoline)
  • PTB approval D- 333-009 618/00
Specially developped for sampling flammable liquids of hazardous materials such as gasoline.
Certifications: PTB approval D- 333-009 618/00.
Delivery information: Supplied complete with a four-meter hose, glass bottle, vacuum pump and grounding cable in transport case.
Documents
Specifikationer
  • Beskrivning:
    UniSampler "Ex"