Molecular model: two-layer DNA, Molymod®
Molecular model of two-layer DNA, 128
Katalog # 761-0107
Marketsource
Leverantör: SPIRING ENTERPRISES
Molecular model: two-layer DNA, Molymod®
Katalog # 761-0107
Leverantör: SPIRING ENTERPRISES
Marketsource
Gå till:
Produktdetaljer och dokument
This set is designed for learning about phosphate structures of purine, pyrimidine and oxyribose, as well as their relationships within DNA.
The two layers can be put on a support and dismantled for storage.
Beställningsinformation: Includes 128 atoms, 150 bonds, a remote bond and a support.
Specifikationer
- Beskrivning:Molecular model of two-layer DNA
- Antal atomer:128