Cartridge burner
Katalog # 451-0046
Leverantör: R L ENTERPRISES
Specifikationer
- Max. temp.:1000 °C
- Beskrivning:Brännare med patron
- Förp.:1
- För:Butane/propane
Specifikationer
Om denna produkt
Brass with replaceable steel plated perforated head. Designed for use with butane/propane mix gas cartridges. This burner can achieve temperatures in excess of 1000 °C.
- Gas cartridge design enables the burner to be detached and re-attached as often as needed, making it easy to store and transport
Typical burning time of the 220 g cartridge is 2¾ hours.