Mini horizontal electrophoresis system, Owl™ B1A EasyCast™
Katalog # 730-0183
Supplier: THERMO OWL SCIENTIFIC
Specifications
- Beskrivning:Owl B1A EasyCast complete system, with buffer exchange port
- Buffertvolym:400 ml
- Gelstorlek, B×L:70×80 mm
- Provkapacitet:5 to 24 samples in 15 minutes at 150 volts
- Enhet, B×L×H:95×160×80 mm
- Förp.:1
Specifications
About this item
The Owl™ B1A EasyCast™ mini gel system is designed to provide flat, even banding patterns and consistent results with minimum effort. This system is ideal for determination of molecular weight ranges of PCR fragments or larger sized DNA molecules. The all-in-one design allows the option of casting and running a gel in the buffer chamber, eliminating the need for additional equipment.
- Two comb slots on the UV transparent gel tray double sample capacity by doubling the number of sample wells
- Heavy duty, one-piece combs eliminate comb assembly
- UVT gel trays are silk screened with a fluorescent ruler for easy measurement of bands
Ordering information: Each system includes buffer chamber, SuperSafe™ lid with attached power leads, gasketed UVT gel tray and two combs (6- and 10-well, double-sided, 1,0/1,5 mm thick).