pH-/mV-/°C-/jonmätare, bordsmodell, inoLab® pH/ION 7320
Supplier: WTW
Certificates
About this item
This two-channel instrument is ideal for precision measurements of pH, ISE or Redox and automatic documentation acc. to GLP/ AQA as required by accredited laboratories. The high resolution measurements can be controlled conveniently and easily via a graphical user interface.
- Five-point calibration for measuring pH, and up to seven-point for measuring ions
- USB interface for fast data transfer in .csv format or via the optional integrated printer
- CMC function for pH and ISE visualises the ideal measuring range and supports correct measuring
- Graphic display with text menu for convenient handling
- Blank value correction, increment methods: known addition, known subtraction, sample addition, sample subtraction
Ordering information: Levereras utan printer (modell med inbyggd printer finns).
Specifications
- Catalog No:
- 662-1685
- 662-1683
- 662-1684
- 662-1686
- Modell:
- pH/ION 7320
- pH/ION 7320
- pH/ION 7320
- pH/ION 7320
- pH – område:
- −2,000...+20,000
- −2,000...+20,000
- −2,000...+20,000
- −2,000...+20,000
- pH – upplösning:
- 0,01 / 0,001
- 0,01 / 0,001
- 0,01 / 0,001
- 0,01 / 0,001
- pH – noggrannhet:
- ±0,005 / ±0,01
- ±0,005 / ±0,01
- ±0,005 / ±0,01
- ±0,005 / ±0,01
- pH – kalibrering:
- MultiCal® automatic calibration (1 - 5 points)
- MultiCal® automatic calibration (1 - 5 points)
- MultiCal® automatic calibration (1 - 5 points)
- MultiCal® automatic calibration (1 - 5 points)
- mV - område:
- ±1200,0 / ±2500
- ±1200,0 / ±2500
- ±1200,0 / ±2500
- ±1200,0 / ±2500
- mV - noggrannhet:
- ±0,3 / ±1
- ±0,3 / ±1
- ±0,3 / ±1
- ±0,3 / ±1
- ISE område:
- 0 - 999999
- 0 - 999999
- 0 - 999999
- 0 - 999999
- ISE upplösning:
- 1, 2 or 3 significant digits
- 1, 2 or 3 significant digits
- 1, 2 or 3 significant digits
- 1, 2 or 3 significant digits
- ISE kalibrering:
- Automatic calibration (1 - 7 points)
- Automatic calibration (1 - 7 points)
- Automatic calibration (1 - 7 points)
- Automatic calibration (1 - 7 points)
- ISE enheter:
- mg/l, mg/kg, mmol/l, ppm, %
- mg/l, mg/kg, mmol/l, ppm, %
- mg/l, mg/kg, mmol/l, ppm, %
- mg/l, mg/kg, mmol/l, ppm, %
- Temperaturområde:
- −5...+105 °C
- −5...+105 °C
- −5...+105 °C
- −5...+105 °C
- Temperature resolution:
- 0,1 °C
- 0,1 °C
- 0,1 °C
- 0,1 °C
- Temperature accuracy:
- ±0,1 °C
- ±0,1 °C
- ±0,1 °C
- ±0,1 °C
- Display:
- Backlit graphic display with CMC
- Backlit graphic display with CMC
- Backlit graphic display with CMC
- Backlit graphic display with CMC
- Certifiering:
- IP 43, GLP
- IP 43, GLP
- IP 43, GLP
- IP 43, GLP
- Minne kapacitet:
- Manual (500), automatic (5000), programmable
- Manual (500), automatic (5000), programmable
- Manual (500), automatic (5000), programmable
- Manual (500), automatic (5000), programmable
- Anslutningar:
- DIN or BNC
- DIN or BNC
- DIN or BNC
- DIN or BNC
- Utgångar:
- USB
- USB
- USB
- USB