Mechanical cell counters
Catalog # 720-0811
Supplier: DENOMINATOR
Specifications
- Mätområde:0 − 999
- Beskrivning:Manual cell counter
- Material:Metal
- Array:8
- Förp.:1
Specifications
About this item
Metal case mechanical cell counters that can count from 0 to 999.
- Anti-slip strips
- Reset button
- Totalizer