Scintillation vials
21,25 ml, 27×51 mm, Ja
Katalog # 548-0255
Leverantör: MURRAY S
Gå till:
Product Details & Documents
These clear scintillation vials with screw cap are made from low potassium glass.
- Complete with fitted foil lined cap
Documents
MURRT006^01_TDS_EN
Datablad
Specifikationer
- Med lock:Ja
- Diam×H:27×51 mm
- Kapacitet:21,25 ml
- Seq:01