Scintillationsburk
Supplier: KARTELL
Certificates
About this item
HDPE, translucent. Compatible with all scintillation counters. A special screw cap ensures leakproof handling.
HDPE, translucent. Compatible with all scintillation counters. A special screw cap ensures leakproof handling.