VWR®, Caps for Culture and Test Tubes
Catalog # 211-0039
Supplier: VWR Collection
Specifications
- Stängningsstorlek:Lock, ventilationstyp
- för rör:12 mm
- Färg:Ofärgad
- Environmentally Preferable:Low Manufacturing Impact,Sustainable Packaging
- Förp.:1.000