Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Rör
Odlingsrör
Culture tubes, Rimless
Culture tubes, Rimless
Culture tubes, Rimless
Leverantör:
Brand
Certifikat
Expandera bilden
Culture tubes, Rimless
Leverantör:
Brand
Om denna produkt
Made of AR-Glas®.
Autoclavable (at 121 °C/2 bar)
Wall thickness 0,6/0,7 mm
Certifications:
DIN EN 285