Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Brickor/Skålar/Burkar
Instrumentbrickor/skålar
Skål
Skål
Skål
Supplier:
P.S. KUNSTSTOFWERKE
Certificates
Expand image
Skål
Supplier:
P.S. KUNSTSTOFWERKE
About this item
Melamin, vit, polerad.
Hög kemikalieresistens
Tål upp till 80 °C
Robust med slät yta för enkel rengöring
Lågprofil.