Degeltång
Catalog # 231-0354
Supplier: NICKEL ELECTRO
Specifications
- Längd:200 mm
- Förp.:1
Specifications
About this item
Steel, non alloy. For easy, safe handling of crucibles made from a wide variety of materials.
- With grooved tips
- Good thermal resistance
- Robust and easy action