Degeltång
Katalog # 231-0353
Leverantör: NICKEL ELECTRO
Specifikationer
- Längd:200 mm
- Förp.:1
Specifikationer
Om denna produkt
Pure nickel (99,6%). With grooved tips. For easy, safe handling of crucibles made from a wide variety of materials.
- Excellent corrosion resistance
- Good thermal and electrical conductivity
- Excellent thermal resistance