Orders LinkContactUsLinkComponent
VWR®, Precisionstermometer
VWR®, Precisionstermometer
VWR®, Precisionstermometer
Supplier:  VWR Collection
Certificates
VWR®, Precisionstermometer
VWR®, Precisionstermometer
6 Options Available Below
Product Details & Documents
Durac Plus precision thermometers are made with permanently fused markings and fractional degrees to provide precision and ensure accuracy.
  • Mercury-free, contain blue organic liquid against lead-free white back glass
  • Individually serialised
Ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical/medical industry, manufacturing, dairies, breweries and distilleries.
Traceable to NIST/DKD. Compliant with ANSI/SAMA Z236.1-1983.
Certifications: Traceable to NIST.
Delivery information: Includes multi-language (English, French, German, Italian, Portuguese and Spanish) statement of accuracy and traceability.
Product Family Options
Product Information
  • Mätområde
    Noggrannhet
    Indelning
    Längd
    Fyllning
    Nedsänkning
    Typ
    Availability
    Price
  • −1...+51 °C
    ±1,0 °C
    0,1 °C
    460 mm
    76 mm
    Zertifiert
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1 675,00 kr
    1 675,00 kr
    1 * 1 Items
       1 675,00 kr
  • −1...+51 °C
    ±0,6 °C
    0,1 °C
    460 mm
    Total
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1 300,00 kr
    1 300,00 kr
    1 * 1 Items
       1 300,00 kr
  • −1...+101 °C
    ±1,0 °C
    0,1 °C
    610 mm
    Röd sprit
    76 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1 775,00 kr
    1 775,00 kr
    1 * 1 Items
       1 775,00 kr
  • −1...+101 °C
    ±0,6 °C
    0,1 °C
    610 mm
    Total
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1 750,00 kr
    1 750,00 kr
    1 * 1 Items
       1 750,00 kr
  • −1...+201 °C
    ±1 °C; ±1,5 °C >150 °C
    0,2 °C
    610 mm
    76 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1 975,00 kr
    1 975,00 kr
    1 * 1 Items
       1 975,00 kr
  • −1...+201 °C
    ±0,7 °C; ±1 °C >100 °C
    0,2 °C
    610 mm
    Total
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 1 925,00 kr
    1 925,00 kr
    1 * 1 Items
       1 925,00 kr