Styva kort för blodgruppsbestämning
Katalog # 763-0279
Leverantör: ELDON CARD
CAS-nummer:
Specifikationer
- Förp.:1 KIT
- Beskrivning:Sats för 50 blodgruppsbestämningar
- Seq:01
Specifikationer
Om denna produkt
The presence or absence of agglutinates will show the blood type. The blood typing kit contains everything to type one person: 1 Eldoncard, 4 Eldonsticks, 1 alcohol swab, 2 safety lancets, 1 result.card. 2 micropipettes are supplied for 50 blood typings. Shelf-life 24 months at 5-37 °C. The foil is used to protect the cards after use.
- Portable and compact
- Blood grouping on location
- Result after just 2 minutes
- Easy to learn, easy to use
- No need for refrigeration and daily controls
- Whole blood can be used down to 5 μl
- No need for separation of red cells and blood plasma