VWR® M40, Transportsvabb
Catalog # 710-0442
Supplier: VWR Collection
Specifications
- Beskrivning:M40-svabb, Amies
- Material:Plastic body
- Transport medium:Amies gel
- Förp.:50
Specifications
About this item
Transportsvabbsystem med plastskaft och syntetisk spets, med Amies agargel.
- Compliant with CLSI (formerly NCCLS) standard M40-A at 21 and 4 °C
- Without charcoal
Helps to improve recovery and survival of fastidious microorganisms from the throat, vagina, wounds and skin swabs.
Certifications: CLSI (formerly NCCLS).
Caution: *Produkten/produkterna på denna sida ligger utanför tillämpningsområdet för EU -direktivet 2019/904