ContactUsLinkComponent
Transport swabs
Transport swabs
Katalog # SLIN190C.USE_
Leverantör:  STERILIN
undefined
Transport swabs
Katalog # SLIN190C.USE_
Leverantör:  STERILIN
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifikationer
  • Beskrivning:
    Transport swabs
  • Seq:
    06
  • Shaft/head:
    Twisted wire/rayon
  • Transport medium:
    Amies gel
  • Lock, färg:
    Blå
  • Förp.:
    500
Specifikationer
Om denna produkt
Designed for sample collection and transport of bacteria, these swabs contain media for the maintenance of bacteria during transport to the laboratory.
  • Viability of bacteria for 24 to 48 hours
  • Pouch seal turning white on opening as a visible sign of tamper evidence
  • Printed with lot number and expiry date and tamper evident sleeve ensuring traceability and sterility
Tested for performance using a wide range of aerobic and anaerobic organisms ensuring adequate recovery together with sterility and other quality assurance tests.
Certifications: Class lla in accordance with Medical Device Directive 93/42/EEC (for transient invasive use).
Packaging: Unique nitrogen flushed aluminium inner bag with laminated film pouch to prevent fromoxidation and dehydration of media.
Caution: *Produkten/produkterna på denna sida ligger utanför tillämpningsområdet för EU -direktivet 2019/904