Swabs, Steriswabs®
Catalog # 720-0142
Supplier: MEDICAL WIRE EQUIPMENT
Specifications
- Beskrivning:Svabbar i märkta rör
- Material:Plast
- Lock, färg:Blå
- Förp.:250
Specifications
About this item
Swabs with 45 mm breakpoint, dry or pre-moistened with sterile diluent.
- Available in labelled tube or peel pouches
- Triple wrapped and radiation sterilised
Designed for effective monitoring of sterility and contamination in clinically clean or sterile work areas.
Caution: *Produkten/produkterna på denna sida ligger utanför tillämpningsområdet för EU -direktivet 2019/904