ContactUsLinkComponent
Adapters, steam distillation head, Quickfit®
Adapters, steam distillation head, Quickfit®
Catalog # BIBBSH7/23/SC
Supplier:  SciLabware
Adapters, steam distillation head, Quickfit®
Catalog # BIBBSH7/23/SC
Supplier:  SciLabware
Specifications
  • Kärna till kondensator:
    19/26 NS
  • Propp till kolv:
    24/29 NS
  • Seq:
    01
  • Förp.:
    5
Specifications
About this item
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
A splash-head with integral stem for the introduction of steam into flasks. Stem is connected to steam supply via detachable screwthread safety connector.