ContactUsLinkComponent
Distilling bridges, Quickfit®
  201-8692
undefined
Distilling bridges, Quickfit®
  201-8692
  • Kärna:
    24/29 NS
  • Förp.:
    1
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
Used in destillation assemblies in which the condenser is assembled in the vertical mode.