Skyddsglasögon, Skyper
Katalog # 111-1233
Leverantör: UVEX
Specifikationer
- Typ:UV-skydd, reptåliga, Optidur 3000 UV
- Färg:Svart
- Seq:03
- Förp.:1
Specifikationer
Om denna produkt
Safety glasses offering an excellent field of vision. Skyper is the standard version, while Skyper s has been developed for slim faces. Skyper s is also available in stylishly transparent colours as uvex Skybrites.
- Five-position temple length adjustment and adjustable lens inclination
- Integrated side shields
- Comfortable fit due to padded ear pieces
- Replaceable lenses
Weight: <30 g
EN 166-168, EN 170, EN 172; 1F
Certifications: EN 166-168, EN 170, EN 172; 1F