Dippers
300 ml, 381 (Handle) mm
Katalog # NICK5072
Leverantör: NICKEL ELECTRO
Gå till:
Product Details & Documents
Stainless steel. With extra-long handles. For milk sample collection and a wide range of other laboratory and industrial applications.
- Excellent chemical resistance
- Excellent thermal resistance
- Smooth surfaces for simple, easy cleaning
Specifikationer
- Kapacitet:300 ml
- Längd:381 (Handle) mm