Kniv
Katalog # SWAN1231
Leverantör: SWANN MORTON
Specifikationer
- Beskrivning:Craft tool starter set, incl. handle with No. 1 and No. 2 blade
- Förp.:10
Specifikationer
Om denna produkt
Carbon steel blade, with plastic handle. The ideal tool for marquetry, modelling, craft, studio work and horticulture.
- Tumskruv används för att byta och hålla kvar blad
- Handtaget av PS är slagtåligt
Three types of replacement blades available as an option
Ordering information: This starter set includes the plastic handle and two blades (one each No. 1 and No. 2 (carded)). Handle and blades are also available separately.