Rack for scintillation vials, SP Bel-Art
Katalog # 218-1290
Leverantör: Bel-Art Products, a Part of SP
Specifikationer
- Hål Ø.:30 mm
- Beskrivning:Rack for scintillation vials
- Förp.:1
Specifikationer
Om denna produkt
PP, autoklaverbar.
- Two handles for safe and easy transport
- Two levels for good sample visibility
Rack for 24 scintillation vials (3×8 format).
W×D×H: 298×127×95 mm.