Robotspetsar för Beckman®
Leverantör: Thermo Fisher Scientific
Certifikat
Om denna produkt
Spetsar av hög kvalitet, testade på de robotar de är avsedda för, för att säkra kompatibiliteten och en bra passform. Garanterat fria från RNas, DNas, DNA, RNA, pyrogen och ATP.
- PP, genomskinliga
- Spetsar med ART® filter: förhindrar korskontaminering mellan proverna; passar för arbete med DNA, RNA och radioaktiva eller smittfarliga prover
- “Low Retention” spetsar: minskar kvarhållning av prov i spetsen, vilket reducerar provförlust och ökar reproducerbarheten
- Sterila spetsar: gammastrålade
Caution: * Adapter 3772 krävs. Tillgänglig endast direkt från Molecular BioProducts
Specifikationer
- Catalog No:
- 736-0038
- 736-0021
- 736-0015
- 736-0077
- 736-0016
- 736-0100
- 736-0075
- 737-2011
- 736-0019
- 736-0099
- 736-0101
- 736-0096
- 736-0001
- 736-0098
- 736-0617
- 736-0618
- 736-0616
- 736-0083
- 736-0017
- 737-2012
- 737-2014
- 736-0090
- 736-0020
- 736-0018
- 736-0086
- 736-0087
- 736-0000
- 736-0094
- 736-0093
- 736-0022
- 736-0013
- 736-0095
- 736-0070
- 736-0097
- 736-0071
- 736-0088
- 736-0089
- 736-0091
- 736-0092
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Miljöcertificering:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)