PCR box/rack
731-1251
: Brand
- Färg:Blandade
- Beskrivning:PCR-låda/rack
- Förp.:5
PP, assorted colours (red, yellow, green, purple, blue).
- Can be stacked without lids
- Able to withstand temperatures from −80 to +121 °C
Suitable for sample preparation, for keeping and storing 0,2 ml single tubes, 8-strips and 12-strips, and 96-well PCR plates.