Accessories for half masks, 7700 Series
Katalog # NORHN06575008
Leverantör: Honeywell Safety
Specifikationer
- Connection type:N series
- Beskrivning:Filter cartridge class P3, material: ABS - fiber glass paper, protection type: particulate
- Förp.:4