Handskar, JACKSON SAFETY* G40 Nitrile Coated
Nitrile foam coated hand specific gloves with seamless knitted nylon liner.
- Excellent dexterity
- Abrasion resistance rating of 4 (EN Standards)
- Excellent wet and dry grip
- Static dissipative in use
- Silicone-free
- Ecofriendly packaging - no separate insert, no printed bags, less waste
Ideal for applications where tactile sensitivity is required, for example in metal manufacturing and automotive and aerospace assembly.
: EN 388 (4131), EN 1149-2; kat. II
- Catalog No:
- 112-0649
- 112-2574
- 112-0650
- 112-2573
- 112-2575
- Modell:
- Nitrile Coated
- Nitrile Coated
- Nitrile Coated
- Nitrile Coated
- Nitrile Coated
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Engångs/Återanvändbar:
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Fodermaterial:
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Yta:
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Mätare:
- 15
- 15
- 15
- 15
- 15
- Kragmodell:
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- EN standard:
- EN 388 (4131), EN 1149-2
- EN 388 (4131), EN 1149-2
- EN 388 (4131), EN 1149-2
- EN 388 (4131), EN 1149-2
- EN 388 (4131), EN 1149-2
- Antistatisk:
- +
- +
- +
- +
- +