Cleanroom gloves, nitrile, sterile, BioClean Excell™
: NITRITEX
These nitrile gloves with beaded cuffs are hand-specific and anatomically shaped.
- Sterile, powder-free, textured surface
- Contain no natural latex proteins, hypoallergenic
- Extended leaching in pure de-ionised water ensures extremely low levels of chlorides and extractable metals
- Nitrile is resistant to a wide range of chemicals
- Excellent anti-static properties
Suitable for use in Class 10, ISO 4 or M2.5 cleanrooms.
Typical particle count ≥0,5 μm: <1200 counts/cm²
: Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003 + A1:2009, EN 374-1:2016 Type B, EN 374-5:2016 Type B and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Sterilized in accordance with ISO 11137-1:2006.
Suitable for contact with all foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003 + A1:2009, EN 374-1:2016 Type B, EN 374-5:2016 Type B and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Sterilized in accordance with ISO 11137-1:2006.
Suitable for contact with all foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
: One cuffed pair per inner plastic wallet, one inner wallet per 'Easy Tear' PE bag; 10 pairs per sealed outer PE bag, 20 outer bags per carton liner, one carton liner per carton (200 pairs).
- Catalog No:
- 112-1340
- 112-1339
- 112-1334
- 112-1324
- 112-1320
- 112-1348
- 112-1347
- 112-1341
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Engångs/Återanvändbar:
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Pudrad/puderfri:
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Renrumsklass:
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- Kragmodell:
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Krage, tjocklek:
- 0,11 mm (4,33 mil)
- 0,11 mm (4,33 mil)
- 0,11 mm (4,33 mil)
- 0,11 mm (4,33 mil)
- 0,11 mm (4,33 mil)
- 0,11 mm (4,33 mil)
- 0,11 mm (4,33 mil)
- 0,11 mm (4,33 mil)
- Finger, tjocklek:
- 0,17 mm (6,69 mil)
- 0,17 mm (6,69 mil)
- 0,17 mm (6,69 mil)
- 0,17 mm (6,69 mil)
- 0,17 mm (6,69 mil)
- 0,17 mm (6,69 mil)
- 0,17 mm (6,69 mil)
- 0,17 mm (6,69 mil)
- Handflata, tjocklek:
- 0,14 mm (5,51 mil)
- 0,14 mm (5,51 mil)
- 0,14 mm (5,51 mil)
- 0,14 mm (5,51 mil)
- 0,14 mm (5,51 mil)
- 0,14 mm (5,51 mil)
- 0,14 mm (5,51 mil)
- 0,14 mm (5,51 mil)
- EN standard:
- EN 455, EN 374 Type B
- EN 455, EN 374 Type B
- EN 455, EN 374 Type B
- EN 455, EN 374 Type B
- EN 455, EN 374 Type B
- EN 455, EN 374 Type B
- EN 455, EN 374 Type B
- EN 455, EN 374 Type B