Filtertratt MicroFunnel™ Plus
About this item
Ideal for monitoring purified water systems maintained at temperatures up to 90 °C (194 °F).
- Multifunctional
- Leakproof lid
- Squeeze separation
- Available in two capacities
Units function as sample container and filter funnel in one which eliminates a sample transfer step and protects critical samples from contamination.
The unique squeeze separation makes membrane retrieval easy. Choose from 0,45 µm GN-6 Metricel® (mixed cellulose ester) membrane used for most applications (not compatible with hot water applications), 0,2 and 0,45 µm Supor® (polyethersulfone) membrane to minimize binding of inhibitory substances, or Metricel® Black (modified polyethersulfone) membrane (not compatible with hot water applications) for better contrast with light colored colonies.
Available in 100 and 300 ml capacity. Graduations marked on outside of cylinder for better visibility and easy measurement. Gamma irradiated. Gamma irradiation dose is 15 to 30 kGy.
MicroFunnel™ Plus AP disposable filter funnels allow for aseptic collection of a sample through the sample port on the lid of the filter funnel without removal of the lid. Optional filter holders are recommended for use when collection hot water samples.
Supor®: Hydrophilic polyethersulfone, white
Ordering information: Filter funnels PALL4829, PALL4807 and PALL4808 are not compatible with hot water applications; for use with ambient temperature samples only. Other disposable filter funnels are available. See MicroFunnel™ Disposable filter funnels for a broad selection of disposable filter funnels, individually bagged for general testing of aqueous solutions. See MicroFunnel™ ST Disposable filter funnels for a convenient and economical choice for sterility testing within isolators. Sentino™ Microbiology pump - designed specifically for MicroFunnel™ filter funnel. Contact your Avantor sales representative for more information.
7 Options Available Below
Product Details & Documents
Ideal for monitoring purified water systems maintained at temperatures up to 90 °C (194 °F).
- Multifunctional
- Leakproof lid
- Squeeze separation
- Available in two capacities
Units function as sample container and filter funnel in one which eliminates a sample transfer step and protects critical samples from contamination.
The unique squeeze separation makes membrane retrieval easy. Choose from 0,45 µm GN-6 Metricel® (mixed cellulose ester) membrane used for most applications (not compatible with hot water applications), 0,2 and 0,45 µm Supor® (polyethersulfone) membrane to minimize binding of inhibitory substances, or Metricel® Black (modified polyethersulfone) membrane (not compatible with hot water applications) for better contrast with light colored colonies.
Available in 100 and 300 ml capacity. Graduations marked on outside of cylinder for better visibility and easy measurement. Gamma irradiated. Gamma irradiation dose is 15 to 30 kGy.
MicroFunnel™ Plus AP disposable filter funnels allow for aseptic collection of a sample through the sample port on the lid of the filter funnel without removal of the lid. Optional filter holders are recommended for use when collection hot water samples.
Supor®: Hydrophilic polyethersulfone, white
Ordering information: Filter funnels PALL4829, PALL4807 and PALL4808 are not compatible with hot water applications; for use with ambient temperature samples only. Other disposable filter funnels are available. See MicroFunnel™ Disposable filter funnels for a broad selection of disposable filter funnels, individually bagged for general testing of aqueous solutions. See MicroFunnel™ ST Disposable filter funnels for a convenient and economical choice for sterility testing within isolators. Sentino™ Microbiology pump - designed specifically for MicroFunnel™ filter funnel. Contact your Avantor sales representative for more information.
Documents
Certificates
Search by Lot NumberMembrane Filter Funnels, MicroFunnel™, PALL Laboratory
Application Notes
Pall Laboratory Microbiology QC Products
Product Literature
PALL4844_TDS_EN
Datasheet
Specifications for Product Family
Specifications
- Catalog No:
- 516-9891
- 516-1238
- 516-9890
- 515-0052
- 515-0055
- 516-1239
- 515-0157
- Effektiv filteryta:
- 13,46 cm²
- 13,46 cm²
- 13,46 cm²
- 13,46 cm²
- 13,46 cm²
- 13,46 cm²
- 13,46 cm²
- Max. temperature:
- 90 °C
- 90 °C
- Ambient
- 90 °C
- 90 °C
- 90 °C
- 90 °C
- Mått:
- 76H×61W mm
- 76H×61W mm
- 76H×61W mm
- 89H×87W mm
- 76H×61W mm
- 89H×87W mm
- 76H×61W mm
Specifications
Product Family Options
Product Information
- BeskrivningKapacitetMaterialMembranPorstorlekAvailabilityPrice
- MicroFunnel™ Plus100 mlPP/PS/PE with PES-membranSupor®, rutnät0,2 µmLog in for availability1 * 50 Items6 900,00 krSpecifications:
More
Specifications
Cat. No.516-9891Effektiv filteryta13,46 cm²Max. temperature90 °CMått76H×61W mm - MicroFunnel™ Plus filter funnel100 mlPESupor® white, gridded0,45 µmLog in for availability1 * 50 Items7 250,00 krSpecifications:
More
Specifications
Cat. No.516-1238Effektiv filteryta13,46 cm²Max. temperature90 °CMått76H×61W mm - MicroFunnel™ Plus100 mlPP/PS/PE with CE-membranMetricel svart, rutnät0,45 µmLog in for availability1 * 50 Items7 000,00 krSpecifications:
More
Specifications
Cat. No.516-9890Effektiv filteryta13,46 cm²Max. temperatureAmbientMått76H×61W mm - MicroFunnel™ Plus300 mlPP/PS/PE with PES-membranSupor®, rutnät0,45 µmLog in for availability1 * 20 Items3 300,00 krSpecifications:
More
Specifications
Cat. No.515-0052Effektiv filteryta13,46 cm²Max. temperature90 °CMått89H×87W mm - MicroFunnel™ Plus100 mlPP/PS/PE with PES-membranSupor®, rutnät0,45 µmLog in for availability1 * 50 Items6 825,00 krSpecifications:
More
Specifications
Cat. No.515-0055Effektiv filteryta13,46 cm²Max. temperature90 °CMått76H×61W mm - MicroFunnel™ Plus filter funnel300 mlPESupor® white, gridded0,45 µmLog in for availability1 * 20 Items3 500,00 krSpecifications:
More
Specifications
Cat. No.516-1239Effektiv filteryta13,46 cm²Max. temperature90 °CMått89H×87W mm - MicroFunnel™ Plus100 mlPP/PS/PE with PES-membranSupor®, rutat (Plus AP)0,45 µmLog in for availability1 * 50 Items7 725,00 krSpecifications:
More
Specifications
Cat. No.515-0157Effektiv filteryta13,46 cm²Max. temperature90 °CMått76H×61W mm
Accessories
Product Information
- Catalog #BeskrivningAvailabilityPrice
- 516-7706Autoklaverbara adaptrar för vakuumgrenrör vid användning av gummiproppLog in for availability1 * 3 Items - 1 025,00 kr
Product Information
- Catalog #BeskrivningAvailabilityPrice
Product Information
- Catalog #BeskrivningAvailabilityPrice
Product Information
- Catalog #BeskrivningAvailabilityPrice
Product Information
- Catalog #BeskrivningAvailabilityPrice
Product Information
- Catalog #BeskrivningAvailabilityPrice
- PALL4863Second generation vacuum pump adapter for MicroFunnel™ filter funnel for alternate pumpLog in for availability1 * 1 Items - 645,00 kr