Needle holder for specimen loops and dissecting needles
Catalog # 631-7142
Supplier: HAMMACHER KARL
Specifications
- Beskrivning:Needle holder
- Längd:170 mm
- Förp.:1
Specifications
About this item
Needle holder for for specimen loops and dissecting needles up to max. diameter of 1,0 mm.