Kolonner, Hamilton PRP-X500
Supplier: HAMILTON BONADUZ
Certificates
About this item
PRP-X500 is a superficially porous polymeric anion exchange column designed for the separation, purification and isolation of proteins, peptides and DNA/RNA. The methacrylate polymeric coating of the PRP-X500 provides a more hydrophilic surface, preventing hydrophobic interaction sample losses.
- Improved mass-transfer, shortened run times and improved resolution
- Fast separations
- Good sample capacity
Specifications
- Catalog No:
- 554-0254
- 554-1315
- 554-1242
- 554-1327
- Max. tryck:
- 5000 psi
- 5000 psi
- 5000 psi
- 5000 psi
- pH område:
- 1 - 13
- 1 - 13
- 1 - 13
- 1 - 13
- Temperature limit:
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
- 5 - 60 °C (pH 1 - 7,9); 5 - 30 °C (pH 8 - 13)
- USP code:
- L23
- L23
- L23
- L23