Cation exchange HPLC columns, HC-75
Supplier: HAMILTON BONADUZ
Certificates
About this item
HC-75 columns separate compounds through ligand exchange. The different forms of the HC-75 (hydrogen, calcium and lead) each provide a unique selectivity for separating varying types of charged analytes based on electronegativity toward the counterion.
- Higher carbohydrate oligomers elute before smaller di- and monosaccharaides
Specifications
- Catalog No:
- 554-1500
- 554-1234
- 554-1233
- 554-0299
- 554-0240
- 554-0235
- 554-2205
- 554-0239
- HAMI79919
- 554-0256
- 554-1303
- 554-2294
- HAMI79917
- Max. tryck:
- 400 psi
- 400 psi
- 400 psi
- 400 psi
- 400 psi
- 400 psi
- 400 psi
- 400 psi
- 400 psi
- 400 psi
- 400 psi
- Temperature limit:
- 25 - 90 °C
- 25 - 90 °C
- 25 - 90 °C
- 25 - 90 °C
- 25 - 90 °C
- 25 - 90 °C
- 25 - 90 °C
- 25 - 90 °C
- 25 - 90 °C
- 25 - 90 °C
- 25 - 90 °C
- USP code:
- L34
- L34
- L34
- L34
- L34
- L34
- L34
- L34
- L34
- L34
- L34