Kemikaliebeständig overall, Spacel Plus 3000 RA/GC/PS, PE
113-8676
- Klädstorlek:3XL
- Outfit applikationer:Chemical protection,Industrial
- Färg:Gul
- Seq:06
- Modell:3000 RA GC PS
- Förp.:10
Disposable, liquidtight overalls made from three-layered PE for excellent protection against liquids (chemicals, oils, hydrocarbons).
- Double adhesive flap and welded zipper
- Excellent abrasion, tear and puncture resistance
- Watertight due to 'Spaseal' welding
- Glued gloves and integrated overboots for an increased safety
- Yellow colour for improved visibility
Tjocklek: 100 µm
Färg: Gul
: Type 3/5, EN 340, EN 1511, Cat. III