Orders LinkContactUsLinkComponent
Sprutflaska, vid hals, Nalgene®
Sprutflaska, vid hals, Nalgene®
Sprutflaska, vid hals, Nalgene®
Sprutflaska, vid hals, Nalgene®
Av genomsynlig LDPE med sprutlock av PP.
  • Hög kemikalieresistens
  • Tål upp till 80 °C
  • Läckagesäker
With wide, very practical filling aperture. Soft, very easy to squeeze.
  • Catalog No:
  • 215-0111
  • 215-0112
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Miljöcertificering:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Product Family Options
  • Kapacitet
    Gänga
  • 500 ml
    53 mm
    1 * 4 Items
    1 * 4 Items - 820,00 kr
    820,00 kr
    1 * 4 Items
       820,00 kr
      Low Manufacturing Impact
    Specifications
    Cat. No.
    215-0111
    Environmentally Preferable
    Low Manufacturing Impact
    Miljöcertificering
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 1000 ml
    63 mm
    1 * 2 Items
    1 * 2 Items - 335,00 kr
    335,00 kr
    1 * 2 Items
       335,00 kr
      Low Manufacturing Impact
    Specifications
    Cat. No.
    215-0112
    Environmentally Preferable
    Low Manufacturing Impact
    Miljöcertificering
    NAR Certificate of Retirement
      Low Manufacturing Impact