Wash bottles, wide neck, for microbiological staining solutions, Azlon®
Katalog # 215-0595
Leverantör: SciLabware
Specifikationer
- Beskrivning:Sats med fem tvättflaskor
- Förp.:5
Specifikationer
Om denna produkt
LDPE, med sprutlock av PP.
- Set of five 250 ml venting wash bottles with DripLok® anti-drip valve
- Printed for Lugols Iodine, Crystal Violet, Gram Colour Decolourising, Gram Iodine and Safranine Solutions
- Printed GHS compliant hazard information where relevant and instructions
Certifications: Azlon® wash bottles meet the material requirements of the U.S. Food and Drug Administration as specified in 21 CFR 177.1520.