Wash bottles, glass
Katalog # 215-6290
Leverantör: Lenz Laborglas GmbH & CO.KG
Specifikationer
- Typ:Komplett tvättflaska
- Beskrivning:Wash bottle complete
- Förp.:1
Specifikationer
Om denna produkt
DURAN®, borosilicate glass clear.
- Very good chemical resistance
- Very good thermal resistance
- Fully interchangeable parts
Universal wash bottle consisting of a 1000 ml capacity flat bottomed flask with standard ground joint sleeve NS 29/32 and spray assembly with standard ground joint cone NS 29/32.