Jars, with twist-off thread
Catalog # 216-3045
Supplier: MUELLER KREMPEL
Specifications
- Ø utv.:100,0 mm
- Gänga:58 mm
- Kapacitet:1000 ml
- Höjd:179,0 mm
- Seq:06
- Förp.:14
Specifications
About this item
Amber glass, without lid.
- Good chemical and physical properties
- For uses without high thermal load
- Matching metal lid available as an separately (see 216-3661 and 216-3663)
Well suited for sampling, storage and transport of powders, pastes and liquids. Amber coloured to protect light-sensitive substances.