Orders LinkContactUsLinkComponent
Dropping bottles, LDPE, Nalgene®
Dropping bottles, LDPE, Nalgene®
Dropping bottles, LDPE, Nalgene®
Dropping bottles, LDPE, Nalgene®
LDPE, genomskinlig, med dropptätning och PP-kåpa.
  • Hög kemikalieresistens, tål upp till 80 °C
  • Fastsatt lock kan inte tappas bort
  • Läckagesäker
Dispenserar en droppe i taget. Lätt att fylla på.
  • Catalog No:
  • 215-7871
  • 215-7872
  • 215-7873
  • 215-7874
  • 215-7875
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Miljöcertificering:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Product Family Options
  • Kapacitet
  • 15 ml
    1 * 12 Items
    1 * 12 Items - 535,00 kr
    535,00 kr
    1 * 12 Items
       535,00 kr
      Low Manufacturing Impact
    Specifications
    Cat. No.
    215-7871
    Environmentally Preferable
    Low Manufacturing Impact
    Miljöcertificering
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 30 ml
    1 * 12 Items
    1 * 12 Items - 710,00 kr
    710,00 kr
    1 * 12 Items
       710,00 kr
      Low Manufacturing Impact
    Specifications
    Cat. No.
    215-7872
    Environmentally Preferable
    Low Manufacturing Impact
    Miljöcertificering
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 60 ml
    1 * 12 Items
    1 * 12 Items - 860,00 kr
    860,00 kr
    1 * 12 Items
       860,00 kr
      Low Manufacturing Impact
    Specifications
    Cat. No.
    215-7873
    Environmentally Preferable
    Low Manufacturing Impact
    Miljöcertificering
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 125 ml
    1 * 12 Items
    1 * 12 Items - 1 025,00 kr
    1 025,00 kr
    1 * 12 Items
       1 025,00 kr
      Low Manufacturing Impact
    Specifications
    Cat. No.
    215-7874
    Environmentally Preferable
    Low Manufacturing Impact
    Miljöcertificering
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 250 ml
    1 * 6 Items
    1 * 6 Items - 650,00 kr
    650,00 kr
    1 * 6 Items
       650,00 kr
      Low Manufacturing Impact
    Specifications
    Cat. No.
    215-7875
    Environmentally Preferable
    Low Manufacturing Impact
    Miljöcertificering
    NAR Certificate of Retirement
      Low Manufacturing Impact