Dubbelmuff
Aluminiumlegering, pulverlackad, 13 mm
Catalog # 241-0370
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
Double bosshead made from powder-coated aluminium alloy.
With nickel plated brass thumb screws.
Specifications
- Material:Aluminiumlegering, pulverlackad
- För stavdiameter:13 mm