Double bosshead
25 mm, Anlöpt gjutjärn
Katalog # 241-2550
Leverantör: Lenz Laborglas
Gå till:
Product Details & Documents
Bosshead made from tempered cast iron. For attaching reaction vessel clamps to support stands.
- Clasping range up to 25 mm
- Fast and easy assembly and disassembly via large brass tommy screws
- Robust, durable
Certifications: DIN 12895
Specifikationer
- Spännvidd:25 mm
- Material:Anlöpt gjutjärn