ContactUsLinkComponent
Beakers, low form, Azlon®
Beakers, low form, Azlon®
Catalog # 213-0138
Supplier:  SciLabware
Supplier #: BWN012
undefined
Beakers, low form, Azlon®
Catalog # 213-0138
Supplier:  SciLabware
Supplier #: BWN012
Log in for availability
Select an Option
32 mm
Select an Option
25 ml
Select an Option
47 mm
Select an Option
02
Select an Option
1
About this item
PTFE, opaque, rigid, non-porous.
  • Excellent chemical resistance - resists nearly all chemicals except fluorine and molten alkali metals
  • Can withstand 260 °C
  • Effective substitutes for platinum containers
Caution: It is NOT advisable to use PTFE beakers on a hotplate.