Murexide for complexometry, Fluka™
Glass bottle, 10 g
Leverantör: Honeywell Chemicals
Murexide for complexometry, Fluka™
Katalog # 33414-10G
Leverantör: Honeywell Chemicals
CAS-nummer: 3051-09-0
Gå till:
Product Details & Documents
Synonymer: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt
- CAS-nummer:3051-09-0
- Formel:C₈H₈N₆O₆
- MDL-nummer:MFCD00012777
- Molecular Weight:284.19 g/mol
- Merck Index:13,06330
- Smält Pt:300 °C
- Förvaringstemperatur:Omgivande
- EINECS:221-266-6
Documents
Certificates
Sök efter LOT-nummerSpecifikationer
Specifikationstestresultat
- Suitability for Complexometry:complying
Specifikationer
- Förpackningstyp:Glass bottle
- Förp.:10 g