Order Entry
Sweden
ContactUsLinkComponent
Methyl blue, powder or crystals, Sigma-Aldrich®
Methyl blue, powder or crystals, Sigma-Aldrich®
Catalog # M6900-50G
CAS Number:  
undefined
Methyl blue, powder or crystals, Sigma-Aldrich®
Catalog # M6900-50G
CAS Number:  

Specifications

  • Förpackningstyp:
    Glass bottle for solids
  • Förp.:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Appearance (Colour):
    Dark Blue to Dark Brown
  • Appearance (Form):
    Powder or Crystals
  • Solubility (Colour)
    1 mg/ml in H2O:
    Dark Blue
  • E1%
    At Lambda Max 594 - 610 nm:
    ≥650
  • Wavelength:
    594 - 610 nm
  • Ratio
    P-15/P+15
    Solvent: 0.005N HCl:
    0.96 - 1.09
  • Recommended Retest Period:
    4 years

Specification Test Results

About this item

 :
Merck Index:13,06055
Melting Pt:≥23 °C
 :M-Clarity™ quality level = MQ200
 :799.81 g/mol
Storage Temperature:Room Temperature
EINECS:249-352-9
CAS Number:28983-56-4
Formula:C₃₇H₂₇N₃Na₂O₉S₃
MDL Number:MFCD00058509